Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H,1H,2H,3H,3H-2-Iodoperfluoroundecan-1-ol ≥97%

Supplier: MATRIX SCIENTIFIC MS

009015-1G 009015-5G
101796-122EA 61.54 USD InStock
101796-122 101796-124
1H,1H,2H,3H,3H-2-Iodoperfluoroundecan-1-ol ≥97%
1H,1H,2H,3H,3H-2-Iodoperfluoroundecan-1-ol
Hazard: Irritant


Formula: C₁₁H₆F₁₇IO
Boiling Pt: 61 °C
MDL Number: MFCD00236072
CAS Number: 38550-45-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty