Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl perfluoro-3,6-dioxaheptanoate ≥98%

Supplier: MATRIX SCIENTIFIC MS

009034-5G 009034-25G
101796-164EA 118.46 USD InStock
101796-164 101796-166
Methyl perfluoro-3,6-dioxaheptanoate ≥98%
Methyl perfluoro-3,6-dioxaheptanoate
Hazard: Irritant


Formula: C₆H₃F₉O₄
MDL Number: MFCD03094140
CAS Number: 39187-41-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty