Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Perfluoro(1,3-dipropoxycyclohexane)

Supplier: MATRIX SCIENTIFIC MS

009100-5G 009100-25G
101796-340EA 112.31 USD InStock
101796-340 101796-342
Perfluoro(1,3-dipropoxycyclohexane)
Perfluoro(1,3-dipropoxycyclohexane)
Hazard: Irritant


Formula: C₁₂F₂₄O₂
MDL Number: MFCD03094119
CAS Number: 400626-83-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty