Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium-4-fluorobenzoate ≥98%

Supplier: MATRIX SCIENTIFIC MS

009243-25G 009243-100G
101796-700EA 43.08 USD InStock
101796-700 101796-702
Sodium-4-fluorobenzoate ≥98%
Sodium-4-fluorobenzoate
Hazard: Irritant


Formula: C₇H₄FO₂Na
MDL Number: MFCD01661384
CAS Number: 499-90-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty