Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Chloromethyl)-3,5-bis(methylsulfonyl)benzene

Supplier: MATRIX SCIENTIFIC MS

009389-250MG 009389-1G
101797-002EA 76.92 USD InStock
101797-002 101797-004
1-(Chloromethyl)-3,5-bis(methylsulfonyl)benzene
1-(Chloromethyl)-3,5-bis(methylsulfonyl)benzene
Hazard: Irritant


Formula: C₉H₁₁ClO₄S₂
MW: 282.77 g/mol
MDL Number: MFCD04037984
CAS Number: 849924-87-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty