Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxy-5-pyrimidineboronic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

009797-1G 009797-5G
101797-286EA 20 USD InStock
101797-286 101797-288
2-Methoxy-5-pyrimidineboronic acid ≥95%
2-Methoxy-5-pyrimidineboronic acid
Hazard: Irritant/Refrigerate


Formula: C₅H₇BN₂O₃
Melting Pt: 159…161 °C
MDL Number: MFCD03094664
CAS Number: 628692-15-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty