Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl dodecafluorosuberate ≥98%

Supplier: MATRIX SCIENTIFIC MS

003150-5G 003150-25G
101785-318EA 307.69 USD InStock
101785-318 101785-320
Dimethyl dodecafluorosuberate ≥98%
Dimethyl dodecafluorosuberate
Hazard: Irritant


Formula: C₁₀H₆F₁₂O₄
MW: 418.14 g/mol
Boiling Pt: 105 °C (1.5 Torr)
MDL Number: MFCD00044088
CAS Number: 2062-20-6
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty