Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H,1H-Perfluoro-n-decylacrylate ≥98%

Supplier: MATRIX SCIENTIFIC MS

003596-5G 003596-25G
101785-906EA 135.38 USD InStock
101785-906 101785-908
1H,1H-Perfluoro-n-decylacrylate ≥98%
1H,1H-Perfluoro-n-decylacrylate
Hazard: Irritant/Refrigerate


Formula: C₁₃H₅F₁₉O₂
Boiling Pt: 220°C/740 Torr
Density: 1.689
MDL Number: MFCD02183510
CAS Number: 335-83-1
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty