Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2-Dibromotrifluoroethyl trifluoromethylether ≥97%

Supplier: MATRIX SCIENTIFIC MS

004600-1G 004600-5G
101787-566EA 44.62 USD InStock
101787-566 101787-568
1,2-Dibromotrifluoroethyl trifluoromethylether ≥97%
1,2-Dibromotrifluoroethyl trifluoromethylether
Hazard: Irritant


Formula: C₃Br₂F₆O
Boiling Pt: 66°C/761 Torr
Density: 2.1096
MDL Number: MFCD01320784
CAS Number: 2356-57-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty