Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-Asp(OtBu)-OH (U-13C4, 15N)

Supplier: MATRIX SCIENTIFIC MS

041300-100MG 041300-250MG
101851-266EA 2544.62 USD InStock
101851-266 101851-268
Fmoc-Asp(OtBu)-OH (U-13C4, 15N)
Fmoc-Asp(OtBu)-OH (U-13C4, 15N)
Hazard: Irritant


Formula: C₂₃H₂₅NO₆
MW: 411.46 g/mol
MDL Number: MFCD09750592
CAS Number: 1217468-27-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty