Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-2-amino-2-indancarboxylic acid

Supplier: MATRIX SCIENTIFIC MS

041380-250MG 041380-1G
101851-544EA 661.54 USD InStock
101851-544 101851-546
Fmoc-2-amino-2-indancarboxylic acid
Fmoc-2-amino-2-indancarboxylic acid
Hazard: Irritant


Formula: C₂₅H₂₁NO₄
MW: 399.45 g/mol
MDL Number: MFCD00800588
CAS Number: 135944-07-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty