Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-L-(3-thienyl)glycine

Supplier: MATRIX SCIENTIFIC MS

041469-100MG 041469-500MG
101851-902EA 598.46 USD InStock
101851-902 101851-904
Boc-L-(3-thienyl)glycine
Boc-L-(3-thienyl)glycine
Hazard: Irritant


Formula: C₁₁H₁₅NO₄S
MW: 257.31 g/mol
MDL Number: MFCD02682479
CAS Number: 910309-12-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty