Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Di-fmoc-N-α-aminomethyl-L-alanine

Supplier: MATRIX SCIENTIFIC MS

041486-50MG 041486-100MG
101851-970EA 1095.38 USD InStock
101851-970 101851-972
Di-fmoc-N-α-aminomethyl-L-alanine
Di-fmoc-N-alpha-aminomethyl-L-alanine
Hazard: Irritant


Formula: C₃₄H₃₀N₂O₆
MW: 562.63 g/mol
MDL Number: MFCD04974553
CAS Number: 1562433-56-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty