Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-(N-β-1-(4,4-dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl)-D-α,β-diaminopropionic acid

Supplier: MATRIX SCIENTIFIC MS

041488-1G 041488-5G
101851-978EA 623.08 USD InStock
101851-978 101851-980
Fmoc-(N-β-1-(4,4-dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl)-D-α,β-diaminopropionic acid
Fmoc-(N-beta-1-(4,4-dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl)-D-alpha,beta-diaminopropionic acid
Hazard: Irritant


Formula: C₂₈H₃₀N₂O₆
MW: 490.56 g/mol
MDL Number: MFCD01631975
CAS Number: 210830-03-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty