Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-5,5,5-trifluoro-DL-leucine

Supplier: MATRIX SCIENTIFIC MS

041512-100MG 041512-500MG
101852-076EA 607.69 USD InStock
101852-076 101852-078
Fmoc-5,5,5-trifluoro-DL-leucine
Fmoc-5,5,5-trifluoro-DL-leucine
Hazard: Irritant


Formula: C₂₁H₂₀F₃NO₄
MDL Number: MFCD02682484
CAS Number: 777946-04-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty