Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-D-(2-thienyl)glycine

Supplier: MATRIX SCIENTIFIC MS

041558-100MG 041558-500MG
101852-262EA 547.69 USD InStock
101852-262 101852-264
Fmoc-D-(2-thienyl)glycine
Fmoc-D-(2-thienyl)glycine
Hazard: Irritant


Formula: C₂₁H₁₇NO₄S
MW: 379.44 g/mol
MDL Number: MFCD07371939
CAS Number: 211682-13-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty