Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-D-propargylglycine ≥95%

Supplier: MATRIX SCIENTIFIC MS

041579-1G 041579-5G
101852-342EA 60 USD InStock
101852-342 101852-344
Fmoc-D-propargylglycine ≥95%
Fmoc-D-propargylglycine
Hazard: Irritant


Formula: C₂₀H₁₇NO₄
MW: 335.359 g/mol
MDL Number: MFCD01311781
CAS Number: 220497-98-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty