Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-α-methyl-DL-glutamic acid

Supplier: MATRIX SCIENTIFIC MS

041646-1G 041646-5G
101852-614EA 735.38 USD InStock
101852-614 101852-616
Fmoc-α-methyl-DL-glutamic acid
Fmoc-alpha-methyl-DL-glutamic acid
Hazard: Irritant


Formula: C₂₁H₂₁NO₆
MW: 383.4 g/mol
MDL Number: MFCD02682512
CAS Number: 1219372-49-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty