Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-D-Gla(otbu)2-OH

Supplier: MATRIX SCIENTIFIC MS

041653-250MG 041653-1G 041653-5G
101852-642EA 958.46 USD InStock
101852-642 101852-644 101852-646
Fmoc-D-Gla(otbu)2-OH
Fmoc-D-Gla(otbu)2-OH
Hazard: Irritant


Formula: C₂₉H₃₅NO₈
MDL Number: MFCD00672341
CAS Number: 111662-65-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty