Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-2-amino-6-methylbenzoic acid

Supplier: MATRIX SCIENTIFIC MS

041680-1G 041680-5G
101852-752EA 364.62 USD InStock
101852-752 101852-754
Boc-2-amino-6-methylbenzoic acid
Boc-2-amino-6-methylbenzoic acid


Formula: C₁₃H₁₇NO₄
MDL Number: MFCD02682223
CAS Number: 269391-47-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty