Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ac-Cys(farnesyl)-OMe

Supplier: MATRIX SCIENTIFIC MS

041741-100MG 041741-500MG
101852-992EA 436.92 USD InStock
101852-992 101852-994
Ac-Cys(farnesyl)-OMe
Ac-Cys(farnesyl)-OMe
Hazard: Irritant


Formula: C₂₁H₃₅NO₃S
MDL Number: MFCD00236994
CAS Number: 135304-08-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty