Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Di-fmoc-seleno-L-cystine

Supplier: MATRIX SCIENTIFIC MS

041786-250MG 041786-1G
101853-182EA 823.08 USD InStock
101853-182 101853-184
Di-fmoc-seleno-L-cystine
Di-fmoc-seleno-L-cystine
Hazard: Irritant


Formula: C₃₆H₃₂N₂O₈Se₂
MDL Number: MFCD04974561
CAS Number: 1639843-33-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty