Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-(2S,3RS)-2-amino-3-hydroxy-4-methylpentanoic acid

Supplier: MATRIX SCIENTIFIC MS

042068-100MG 042068-250MG
101854-326EA 400 USD InStock
101854-326 101854-328
Boc-(2S,3RS)-2-amino-3-hydroxy-4-methylpentanoic acid
Boc-(2S,3RS)-2-amino-3-hydroxy-4-methylpentanoic acid
Hazard: Irritant


Formula: C₁₁H₂₁NO₅
MDL Number: MFCD02682586
CAS Number: 1217531-78-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty