Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-(R)-2-amino-3-hydroxy-3-methylbutanoic acid

Supplier: MATRIX SCIENTIFIC MS

042098-100MG 042098-500MG
101854-460EA 412.31 USD InStock
101854-460 101854-462
Fmoc-(R)-2-amino-3-hydroxy-3-methylbutanoic acid
Fmoc-(R)-2-amino-3-hydroxy-3-methylbutanoic acid
Hazard: Irritant


Formula: C₂₀H₂₁NO₅
MW: 355.39 g/mol
MDL Number: MFCD02682577
CAS Number: 884880-39-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty