Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-(S)-2-amino-3-ethoxypropionic acid

Supplier: MATRIX SCIENTIFIC MS

042101-1G 042101-5G
101854-472EA 465.58 USD InStock
101854-472 101854-474
Fmoc-(S)-2-amino-3-ethoxypropionic acid
Fmoc-(S)-2-amino-3-ethoxypropionic acid
Hazard: Irritant


Formula: C₂₀H₂₁NO₅
MDL Number: MFCD09750517
CAS Number: 1562432-09-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty