Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-D-α-methylserine

Supplier: MATRIX SCIENTIFIC MS

042109-100MG 042109-1G
101854-506EA 323.08 USD InStock
101854-506 101854-508
Fmoc-D-α-methylserine
Fmoc-D-alpha-methylserine
Hazard: Irritant


Formula: C₁₉H₁₉NO₅
MW: 341.37 g/mol
MDL Number: MFCD02682598
CAS Number: 1217644-25-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty