Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-7-benzyloxy-DL-tryptophan

Supplier: MATRIX SCIENTIFIC MS

042129-10MG 042129-100MG
101854-586EA 598.46 USD InStock
101854-586 101854-588
Boc-7-benzyloxy-DL-tryptophan
Boc-7-benzyloxy-DL-tryptophan
Hazard: Irritant


Formula: C₂₃H₂₆N₂O₅
MW: 410.47 g/mol
MDL Number: MFCD02682354
CAS Number: 1219375-22-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty