Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-4-aminooxanilic acid

Supplier: MATRIX SCIENTIFIC MS

042196-500MG 042196-1G
101854-850EA 436.92 USD InStock
101854-850 101854-852
Fmoc-4-aminooxanilic acid
Fmoc-4-aminooxanilic acid
Hazard: Irritant


Formula: C₂₃H₁₈N₂O₅
MW: 402.41 g/mol
MDL Number: MFCD12547720
CAS Number: 186320-17-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty