Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Propoxycinnamic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

047589-500MG 047589-1G 047589-5G
101880-688EA 181.54 USD InStock
101880-688 101880-690 101880-692
4-Propoxycinnamic acid ≥95%
4-Propoxycinnamic acid
Hazard: Irritant


Formula: C₁₂H₁₄O₃
MDL Number: MFCD00995149
CAS Number: 69033-81-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty