Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclobutylmethyl-3-piperidinylmethyl ether hydrochloride

Supplier: MATRIX SCIENTIFIC MS

050915-500MG 050915-2.5G
101893-552EA 364.62 USD InStock
101893-552 101893-554
Cyclobutylmethyl-3-piperidinylmethyl ether hydrochloride
Cyclobutylmethyl-3-piperidinylmethyl ether hydrochloride
Hazard: Irritant


Formula: C₁₁H₂₂ClNO
MDL Number: MFCD13561285
CAS Number: 1211505-35-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty