Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl (2S,4S)-4-[(5-chloro[1,1'-biphenyl]-2-yl)oxy]-2-pyrrolidinecarboxylate hydrochloride

Supplier: MATRIX SCIENTIFIC MS

050994-500MG 050994-2.5G
101893-868EA 364.62 USD InStock
101893-868 101893-870
Methyl (2S,4S)-4-[(5-chloro[1,1'-biphenyl]-2-yl)oxy]-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-[(5-chloro[1,1'-biphenyl]-2-yl)oxy]-2-pyrrolidinecarboxylate hydrochloride
Hazard: Irritant


Formula: C₁₈H₁₉Cl₂NO₃
MDL Number: MFCD13561364
CAS Number: 1354486-26-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty