Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Fluoro-6-(pyrrolidin-1-yl)pyridin-3-yl)ethanone

Supplier: MATRIX SCIENTIFIC MS

054108-100MG 054108-500MG
101906-198EA 364.62 USD InStock
101906-198 101906-200
1-(2-Fluoro-6-(pyrrolidin-1-yl)pyridin-3-yl)ethanone
1-(2-Fluoro-6-(pyrrolidin-1-yl)pyridin-3-yl)ethanone
Hazard: Irritant


Formula: C₁₁H₁₃FN₂O
MW: 208.24 g/mol
MDL Number: MFCD13563070
CAS Number: 1203499-51-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty