Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Racemic trans)-1-tert-Butyl-3-methyl-4-(2-chloro-5-methylpyridin-3-yl)pyrrolidine-1,3-dicarboxylate

Supplier: MATRIX SCIENTIFIC MS

054116-100MG 054116-500MG
101906-230EA 364.62 USD InStock
101906-230 101906-232
(Racemic trans)-1-tert-Butyl-3-methyl-4-(2-chloro-5-methylpyridin-3-yl)pyrrolidine-1,3-dicarboxylate
(Racemic trans)-1-tert-Butyl-3-methyl-4-(2-chloro-5-methylpyridin-3-yl)pyrrolidine-1,3-dicarboxylate
Hazard: Irritant


Formula: C₁₇H₂₃ClN₂O₄
MDL Number: MFCD13563077
CAS Number: 1228070-72-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty