Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1H-Indol-3-ylsulfanyl)methanimidamide hydroiodide ≥95%

Supplier: MATRIX SCIENTIFIC MS

054191-500MG 054191-1G 054191-5G
101906-516EA 181.54 USD InStock
101906-516 101906-518 101906-520
(1H-Indol-3-ylsulfanyl)methanimidamide hydroiodide ≥95%
(1H-Indol-3-ylsulfanyl)methanimidamide hydroiodide
Hazard: Irritant


Formula: C₉H₁₀IN₃S
MDL Number: MFCD00012688
CAS Number: 26377-76-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty