Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-3-(trifluoromethyl)-2-pyridine carboxylic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

054280-500MG 054280-1G 054280-5G
101906-888EA 846.15 USD InStock
101906-888 101906-890 101906-892
5-Chloro-3-(trifluoromethyl)-2-pyridine carboxylic acid ≥95%
5-Chloro-3-(trifluoromethyl)-2-pyridine carboxylic acid
Hazard: Irritant


Formula: C₇H₃ClF₃NO₂
MW: 225.56 g/mol
MDL Number: MFCD14155643
CAS Number: 1198475-43-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty