Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2H,4H,5H,6H,7H-Pyrazolo[4,3-c]pyridine dihydrochloride ≥95%

Supplier: MATRIX SCIENTIFIC MS

054289-500MG 054289-1G 054289-5G
101906-930EA 1521.54 USD InStock
101906-930 101906-932 101906-934
2H,4H,5H,6H,7H-Pyrazolo[4,3-c]pyridine dihydrochloride ≥95%
2H,4H,5H,6H,7H-Pyrazolo[4,3-c]pyridine dihydrochloride
Hazard: Irritant


Formula: C₆H₁₁Cl₂N₃
MDL Number: MFCD13368261
CAS Number: 410544-19-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty