Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-methoxy-5-methyl-3-nitrobenzenecarboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

054300-500MG 054300-1G 054300-5G
101906-974EA 556.92 USD InStock
101906-974 101906-976 101906-978
Methyl-2-methoxy-5-methyl-3-nitrobenzenecarboxylate ≥95%
Methyl-2-methoxy-5-methyl-3-nitrobenzenecarboxylate
Hazard: Irritant


Formula: C₁₀H₁₁NO₅
MW: 225.2 g/mol
MDL Number: MFCD14560554
CAS Number: 863226-26-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty