Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-6-bromo-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-8-carboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

054301-500MG 054301-1G
101906-980EA 1109.23 USD InStock
101906-980 101906-982
Methyl-6-bromo-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-8-carboxylate ≥95%
Methyl-6-bromo-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-8-carboxylate
Hazard: Irritant


Formula: C₁₀H₈BrNO₄
MDL Number: MFCD12196467
CAS Number: 141761-85-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty