Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-[2-(4-benzylpiperazino)ethoxy]benzenecarboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

054315-500MG 054315-1G 054315-5G
101907-032EA 278.46 USD InStock
101907-032 101907-034 101907-036
Methyl-3-[2-(4-benzylpiperazino)ethoxy]benzenecarboxylate ≥95%
Methyl-3-[2-(4-benzylpiperazino)ethoxy]benzenecarboxylate
Hazard: Irritant


Formula: C₂₁H₂₆N₂O₃
MDL Number: MFCD14581671
CAS Number: 1203898-15-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty