Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Amino-N,N-dimethylpentanamide oxalic acid salt ≥95%

Supplier: MATRIX SCIENTIFIC MS

054669-500MG 054669-1G
101908-062EA 921.54 USD InStock
101908-062 101908-064
5-Amino-N,N-dimethylpentanamide oxalic acid salt ≥95%
5-Amino-N,N-dimethylpentanamide oxalic acid salt
Hazard: Irritant


Formula: C₉H₁₈N₂O₅
MDL Number: MFCD16090035
CAS Number: 1221792-33-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty