Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-1,2,3,4-tetrahydronorharman-3-carboxylic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

054764-500MG 054764-1G
101908-434EA 921.54 USD InStock
101908-434 101908-436
Fmoc-1,2,3,4-tetrahydronorharman-3-carboxylic acid ≥95%
Fmoc-1,2,3,4-tetrahydronorharman-3-carboxylic acid
Hazard: Irritant


Formula: C₂₇H₂₂N₂O₄
MDL Number: MFCD03453207
CAS Number: 1122062-52-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty