Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-7-fluoro-5H-pyrrolo[3,2-d]pyrimidine ≥96%

Supplier: MATRIX SCIENTIFIC MS

059872-100MG 059872-250MG
101919-366EA 218.46 USD InStock
101919-366 101919-368
4-Chloro-7-fluoro-5H-pyrrolo[3,2-d]pyrimidine ≥96%
4-Chloro-7-fluoro-5H-pyrrolo[3,2-d]pyrimidine
Hazard: Irritant


Formula: C₆H₃ClFN₃
MW: 171.56 g/mol
MDL Number: MFCD19105251
CAS Number: 1311275-30-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty