Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl thiazole-5-carboxylate ≥98%

Supplier: MATRIX SCIENTIFIC MS

069452-1G 069452-5G 069452-25G
101940-200EA 698.46 USD InStock
101940-200 101940-202 101940-204
Methyl thiazole-5-carboxylate ≥98%
Methyl thiazole-5-carboxylate
Hazard: Irritant


Formula: C₅H₅NO₂S
MDL Number: MFCD03788563
CAS Number: 14527-44-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty