Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Methylthio)-5-pyridineboronic acid ≥98%

Supplier: MATRIX SCIENTIFIC MS

069538-1G 069538-5G 069538-25G
101940-684EA 349.23 USD InStock
101940-684 101940-686 101940-688
2-(Methylthio)-5-pyridineboronic acid ≥98%
2-(Methylthio)-5-pyridineboronic acid
Hazard: Irritant


Formula: C₆H₈BNO₂S
MDL Number: MFCD03788238
CAS Number: 321438-86-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty