Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2-Difluorobenzo[1,3]dioxole-4-boronic acid ≥97%

Supplier: MATRIX SCIENTIFIC MS

069573-500MG 069573-1G 069573-5G
101940-874EA 384.62 USD InStock
101940-874 101940-876 101940-878
2,2-Difluorobenzo[1,3]dioxole-4-boronic acid ≥97%
2,2-Difluorobenzo[1,3]dioxole-4-boronic acid
Hazard: Irritant


Formula: C₇H₅BF₂O₄
MDL Number: MFCD08062343
CAS Number: 126120-87-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty