Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Pyrazinecarbonyl chloride

Supplier: MATRIX SCIENTIFIC MS

069603-1G 069603-5G 069603-25G
101941-030EA 349.23 USD InStock
101941-030 101941-032 101941-034
2-Pyrazinecarbonyl chloride
2-Pyrazinecarbonyl chloride
Hazard: Irritant


Formula: C₅H₃ClN₂O
MDL Number: MFCD00792886
CAS Number: 19847-10-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty