Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-2H-indazole-7-carboxylic acid methyl ester ≥97%

Supplier: MATRIX SCIENTIFIC MS

069843-250MG 069843-1G
101942-192EA 466.15 USD InStock
101942-192 101942-196
2-Methyl-2H-indazole-7-carboxylic acid methyl ester ≥97%
2-Methyl-2H-indazole-7-carboxylic acid methyl ester
Hazard: Irritant


Formula: C₁₀H₁₀N₂O₂
MDL Number: MFCD11109408
CAS Number: 1092351-88-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty