Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5,6,7,8-tetrahydro-1,6-naphthyridine hydrochloride ≥95%

Supplier: MATRIX SCIENTIFIC MS

069956-5G 069956-25G
101942-694EA 315.38 USD InStock
101942-694 102551-762
2-Chloro-5,6,7,8-tetrahydro-1,6-naphthyridine hydrochloride ≥95%
2-Chloro-5,6,7,8-tetrahydro-1,6-naphthyridine hydrochloride
Hazard: Irritant


Formula: C₈H₁₀Cl₂N₂
MDL Number: MFCD09835503
CAS Number: 766545-20-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty