Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Methyl-1H-indazole-5-boronic acid ≥96%

Supplier: MATRIX SCIENTIFIC MS

070140-500MG 070140-1G 070140-5G
101943-500EA 510.77 USD InStock
101943-500 101943-502 101943-504
7-Methyl-1H-indazole-5-boronic acid ≥96%
7-Methyl-1H-indazole-5-boronic acid
Hazard: Irritant/Store in Freezer


Formula: C₈H₉BN₂O₂
MDL Number: MFCD12028646
CAS Number: 1245816-07-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty