Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-5-(aminomethyl)-1,3,4-oxadiazol-2-carboxylate trifluoroacetic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

070215-1G 070215-5G
101943-816EA 1570.77 USD InStock
101943-816 101943-818
Ethyl-5-(aminomethyl)-1,3,4-oxadiazol-2-carboxylate trifluoroacetic acid ≥95%
Ethyl-5-(aminomethyl)-1,3,4-oxadiazol-2-carboxylate trifluoroacetic acid
Hazard: Irritant


Formula: C₈H₁₀F₃N₃O₅
MW: 285.18 g/mol
MDL Number: MFCD11109486
CAS Number: 751479-66-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty